Triklorättiksyra (frostad) 99
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: TCA
A11156.0B A11156.0I A11156.30 A11156.36
A11156.0BEA 2900 SEK
A11156.0B A11156.0I A11156.30 A11156.36
Triklorättiksyra (frostad) 99
Triklorättiksyra (frostad)
Purity: 99% CAS No.: 76-03-9 Molecular Formula: Cl3CCO2H Formula Weight: 163.39 Odour: Acrid pH: 1 Melting Point: 54-58° Boiling Point: 196° Beilstein Reference: 970119 Merck Reference: 14,9627 UN No.: 1839 MDL No.: MFCD00004177
Formel: Cl₃CCOOH
MW: 163.39 g/mol
Kokpunkt: 196 °C
Smältpunkt: 54…58 °C
Densitet: 1.629
Flampunkt: 113 °C (closed cup)
Storage Temperature: Omgivande
MDL Number: MFCD00004177
CAS nummer: 76-03-9
EINECS: 200-927-2
ADR: 8 II
Merck Index: 13,09700
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae34130684af3f73eccad44eac686e325855232d&vid=b600bf2ba67ce275263e6172675e131d3f263837&prid=web&clid=SAPDAM