Diklormetan ≥99% stabilised
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Metylenklorid
Stabilization: 50 ppm Stabiliseras med 2-metyl-2-buten
L13089.AP L13089.AU L13089.0F
L13089.APEA 615 SEK
L13089.AP L13089.AU L13089.0F
Diklormetan ≥99% stabilised
Diklormetan
Stabilized with ca. 50 ppm amylene
Purity: 99+% CAS No.: 75-09-2 Molecular Formula: CH2Cl2 Formula Weight: 84.93 Refractive Index: 1.4244 Odour: Sweet Boiling Point: 39-40° Beilstein Reference: 1730800 Merck Reference: 14,6063 UN No.: 1593 MDL No.: MFCD00000881
Formel: CH₂Cl₂
MW: 84.93 g/mol
Kokpunkt: 39…40 °C
Smältpunkt: –95 °C
Densitet: 1.325
Storage Temperature: Omgivande
MDL Number: MFCD00000881
CAS nummer: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1 III
Merck Index: 14,6063
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM