1-Klorbutan 99+%, ren
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: n-Butyl Chloride
154630010 154630025 154630100
ACRO154630010EA 940 SEK
ACRO154630010 ACRO154630025 ACRO154630100
1-Klorbutan 99+%, ren
1-Klorbutan
Formel: CH₃(CH₂)₃Cl
MW: 92.57 g/mol
Kokpunkt: 77…78 °C (760 mmHg)
Smältpunkt: –123 °C
Densitet: 0.88 g/cm³
Flampunkt: –12 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001009
CAS nummer: 109-69-3
EINECS: 203-696-6
UN: 1127
ADR: 3 II
Merck Index: 13,01559
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2bd980eb6e7ecd91951d46d0ea510119e976dc0&vid=5231549172a609ad96623a80204b11c7c0c69273&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4005 to 1.4035 (20°C, 589 nm)
GC ≥99.0 %
NMR (1H-NMR) Consistent with structure