1,2-Dichlorobutane ≥97%
Leverantör: Thermo Scientific
Varning
B21307.03 B21307.06
B21307.03EA 815 SEK
B21307.03 B21307.06
1,2-Dichlorobutane ≥97%
1,2-Dichlorobutane
Purity: 97+% CAS No.: 616-21-7 Molecular Formula: C4H8Cl2 Formula Weight: 127.01 Flash Point: 30°(86°F) Refractive Index: 1.4450 Boiling Point: 124° Beilstein Reference: 1731376 UN No.: 1993 MDL No.: MFCD00013676
Formel: ClH₂CCHClCH₂CH₃
MW: 127,01 g/mol
Kokpunkt: 124 °C
Densitet: 1,112
Flampunkt: 30 °C (86 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00013676
CAS nummer: 616-21-7
EINECS: 210-469-5
UN: 1993
ADR: 3,III