Bariumacetat 99,999 Metaller basis, Puratronic®
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Barium di(acetate), Acetic acid barium salt
11126.06 11126.14 11126.22
11126.06EA 2025 SEK
11126.06 11126.14 11126.22
Bariumacetat 99,999 Metaller basis, Puratronic®
Bariumacetat
Purity: 99.999% CAS No.: 543-80-6 Molecular Formula: C4H6BaO4 Formula Weight: 255.43 Physical Form: Crystalline Density: 2.47 Appearance: White Odour: Odourless Beilstein Reference: 3693411 Merck Reference: 14,965 UN No.: 1564 MDL No.: MFCD00012447
Formel: (H₃CCOO)₂Ba
MW: 255.42 g/mol
Smältpunkt: ∼450 °C
Densitet: 2.47
Storage Temperature: Omgivande
MDL Number: MFCD00012447
CAS nummer: 543-80-6
EINECS: 208-849-0
UN: 1564
ADR: 6.1 III
Merck Index: 14,965