Natriumpropionat 99,0-100,5
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Propionic acid sodium salt, Sodiumpropionate
149010010 149010250 149010050
ACRO149010010EA 1050 SEK
ACRO149010010 ACRO149010250 ACRO149010050
Natriumpropionat 99,0-100,5
Natriumpropionat
Formel: C₃H₅NaO₂
MW: 96.06 g/mol
Smältpunkt: 285…286 °C
Flampunkt: >250 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002759
CAS nummer: 137-40-6
EINECS: 205-290-4
Merck Index: 12,08816
Order Now
Specifikation testresultat
Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤1 % (K.F.)
Titration with HClO4 99.0 to 100.5 % (On dried substance)
Iron (Fe) ≤30 ppm
Heavy metals (as Pb) ≤10 ppm