Undecanoic acid 99%
Leverantör: Thermo Fisher Scientific
Varning
173970250 173971000 173970050
ACRO173970250EA 655 SEK
ACRO173970250 ACRO173971000 ACRO173970050
Undecanoic acid 99%
Undecanoic acid
Formel: C₁₁H₂₂O₂
Kokpunkt: 228 °C (1013 hPa)
Densitet: 0,89 g/cm³ (20 °C)
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002730
CAS nummer: 112-37-8
EINECS: 203-964-2
Order Now
Specifikation testresultat
Appearance White to light yellow Low melting solid
Infrared spectrum Conforms
GC ≥98.5 %
Titration with NaOH ≥98.5 %