Oktansyra ≥99.5%, liquid
Leverantör: MP Biomedicals
Fara
Synonyms: Octanoic acid, n-Octanoic Acid, Octoic acid
0210039191 0210039125 0210039190
ICNA0210039191EA 3900 SEK
ICNA0210039191 ICNA0210039125 ICNA0210039190
Oktansyra ≥99.5%, liquid
Oktansyra
Caprylic acid is the common name for the eight-carbon saturated fatty acid known by the systematic name octanoic acid. It is found naturally in the milk of various mammals, and it is a minor constituent of coconut oil and palm kernel oil.
Caprylic acid is an intermediate in the manufacture of perfume esters and of dyes.
It is the common name for the eight-carbon saturated fatty acid known by the systematic name octanoic acid. It is found naturally in the milk of various mammals, and it is a minor constituent of coconut oil and palm kernel oil.
Formel: C₈H₁₆O₂
MW: 144,21 g/mol
Kokpunkt: 237 °C (lit.)
Smältpunkt: 15…17 °C (lit.)
Densitet: 0,91 g/cm³ (20 °C) (lit.)
Flampunkt: 130 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004429
CAS nummer: 124-07-2
EINECS: 204-677-5
UN: 3265
ADR: 8,III
Merck Index: 13,01771
Order Now
Specifikation testresultat
Identity Test Passes
Purity 99.6%
FTIR Conforms to Standard