Propylacetat 99
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Acetic acid propyl ester
L15355.AP L15355.AU
L15355.APEA 625 SEK
L15355.AP L15355.AU
Propylacetat 99
Propylacetat
Purity: 99% CAS No.: 109-60-4 Molecular Formula: C5H10O2 Formula Weight: 102.13 Flash Point: 14°(57°F) Refractive Index: 1.3840 Odour: Fruit-like Melting Point: -92° Boiling Point: 99-102° Beilstein Reference: 1740764 Merck Reference: 14,7841 UN No.: 1276 MDL No.: MFCD00009372
Formel: C₅H₁₀O₂
MW: 102.13 g/mol
Kokpunkt: 99…102 °C
Smältpunkt: –92 °C
Densitet: 0.887
Flampunkt: 14 °C (57 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00009372
CAS nummer: 109-60-4
EINECS: 203-686-1
UN: 1276
ADR: 3 II
Merck Index: 12,08026