Pentylacetat 99%
Leverantör: Thermo Scientific
Varning
Synonyms: n-Pentyl acetate, Acetic acid amyl ester, Acetic acid pentyl ester, Pentyl acetate
149180010 149182500 149180025
ACRO149180010EA 1750 SEK
ACRO149180010 ACRO149182500 ACRO149180025
Pentylacetat 99%
Pentylacetat
Formel: C₇H₁₄O₂
MW: 130,19 g/mol
Kokpunkt: 149 °C (760 mmHg)
Smältpunkt: –70,8 °C
Densitet: 0,87 g/cm³
Flampunkt: 24 °C
Storage Temperature: Omgivande
MDL Number: MFCD00009500
CAS nummer: 628-63-7
EINECS: 211-047-3
UN: 1104
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=37af9fce9b4d1452153c5c88d6e47d0d5e08f221&vid=402af1833dd06c8e1652722d88600d04d2775130&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4010 to 1.4030 (20°C, 589 nm)
GC ≥98.5 %
TLC ≥97.5 %
Specific optical rotation -10.8° to -11.8° (20°C, 589 nm) (c=2 in DMF)