Hexyl acetate ≥99%
Leverantör: Thermo Scientific
Varning
Synonyms: Acetic acid hexyl ester
L06342.AK L06342.AU
L06342.AKEA 620 SEK
L06342.AK L06342.AU
Hexyl acetate ≥99%
Hexyl acetate
Purity: 99% CAS No.: 142-92-7 Molecular Formula: C8H16O2 Formula Weight: 144.21 Flash Point: 43°(109°F) Refractive Index: 1.4090 Melting Point: -80° Boiling Point: 167-169° Beilstein Reference: 1747138 UN No.: 3271 MDL No.: MFCD00009524
Formel: C₈H₁₆O₂
MW: 144,21 g/mol
Kokpunkt: 167…169 °C
Smältpunkt: –80 °C
Densitet: 0,873
Flampunkt: 43 °C (109 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00009524
CAS nummer: 142-92-7
EINECS: 205-572-7
UN: 3271
ADR: 3,III