1,3-Dihydroxyaceton, extra pure for cosmetic
Leverantör: Merck
Synonyms: Dihydroxyacetone
1.10150.1000 1.10150.9026
1.10150.1000EA 8580 SEK
1.10150.1000 1.10150.9026
1,3-Dihydroxyaceton, extra pure for cosmetic
1,3-Dihydroxyaceton
Formel: C₃H₆O₃
Smältpunkt: 75 °C
Densitet: 1,52 g/cm³ (20 °C)
Storage Temperature: Kylskåp
MDL Number: MFCD00004670
CAS nummer: 96-26-4
EINECS: 202-494-5