Methoxyacetone 96%

Leverantör: Thermo Scientific
Varning

Synonyms: Methoxy-2-propanone

175730100 175730500
ACRO175730100EA 2300 SEK
ACRO175730100 ACRO175730500
Methoxyacetone 96%
Methoxyacetone
Formel: C₄H₈O₂
MW: 88,11 g/mol
Kokpunkt: 118 °C (1013 hPa)
Smältpunkt: –66 °C
Densitet: 0,95 g/cm³
Flampunkt: 25 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008768
CAS nummer: 5878-19-3
EINECS: 227-549-0
UN: 1993
ADR: 3,III

Order Now

Specifikation testresultat

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3970 to 1.4000 (20°C, 589 nm)
GC ≥95 %
Specific optical rotation -63.5° to -67.5° (20°C, 589 nm) (c=1, water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR