Aceton, Supelco®
Leverantör: Merck
Fara
Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone
M-Clarity™ quality level = MQ100
90872-10ML-F 90872-5ML-F
90872-10ML-FEA 4400 SEK
90872-10ML-F 90872-5ML-F
Aceton, Supelco®
Aceton
Formel: CH₃COCH₃
MW: 58.08 g/mol
Kokpunkt: 56,2
Smältpunkt: –95.4 °C
Densitet: 0.792 g/cm³ (20 °C)
Flampunkt: <–20 °C (closed cup)
Storage Temperature: Omgivande
MDL Number: MFCD00008765
CAS nummer: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3 II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ffd63f2c762ab27d09700d680bfa3a594ad0786&vid=8e311364262fce69c25e7ed63a9c0b8106150cd7&prid=web&clid=SAPDAM