Aceton, EMPROVE® ESSENTIAL NF, BP, Ph. Eur., JP, extra pure suitable for use as excipient, SAFC®
Leverantör: MERCK PRODUCTION CHEMICALS
Fara
Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone
M-Clarity™ quality level = MQ200
1.00013.1000 1.00013.2500 1.00013.6010 1.00013.6025 1.00013.6190 1.00013.9950 1.00013.9180 1.00013.9911
1.00013.1000EA 2200 SEK
1.00013.1000 1.00013.2500 1.00013.6010 1.00013.6025 1.00013.6190 1.00013.9950 1.00013.9180 1.00013.9911
Aceton, EMPROVE® ESSENTIAL NF, BP, Ph. Eur., JP, extra pure suitable for use as excipient, SAFC®
Aceton
Formel: CH₃COCH₃
Kokpunkt: 56,2 °C (1013 hPa)
Densitet: 0,792 g/cm³ (20 °C)
Flampunkt: <–20 °C (closed cup)
Storage Temperature: Omgivande
MDL Number: MFCD00008765
CAS nummer: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM