Aceton for residue analysis
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone
326570010 326570025
ACRO326570010EA 645 SEK
ACRO326570010 ACRO326570025
Aceton for residue analysis
Aceton
Formel: CH₃COCH₃
Kokpunkt: 56,2 °C (1013 hPa)
Smältpunkt: –95 °C
Densitet: 0,79 g/cm³
Flampunkt: –20 °C (closed cup)
Storage Temperature: Omgivande
MDL Number: MFCD00008765
CAS nummer: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Water ≤0.2 %
GC (ECD) Passes test, (NPD) Passes test, (FID) Passes test
Residue after evaporation ≤2 ppm