2-Pentanon ≥99%
Leverantör: Thermo Fisher Scientific
Fara
L13262.14 L13262.22 L13262.36
L13262.14EA 660 SEK
L13262.14 L13262.22 L13262.36
2-Pentanon ≥99%
2-Pentanon
Purity: 99+% CAS No.: 107-87-9 Molecular Formula: C5H10O Formula Weight: 86.13 Flash Point: 7°(44°F) Refractive Index: 1.3905 Melting Point: -78° Boiling Point: 100-101° Beilstein Reference: 506058 Merck Reference: 14,6114 UN No.: 1249 MDL No.: MFCD00009400
Formel: C₅H₁₀O
Kokpunkt: 100…101 °C
Smältpunkt: –78 °C
Densitet: 0,813
Flampunkt: 7 °C (44 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00009400
CAS nummer: 107-87-9
EINECS: 203-528-1
UN: 1249
ADR: 3,II
Merck Index: 14,6114