trans-2-Methyl-2-butenal 98%
Leverantör: Thermo Scientific
Fara
Synonyms: Trans-2-methyl-2-butenal, Tiglic aldehyde, (E)-2-Methyl-2-butenal
353030050 353030250
ACRO353030050EA 630 SEK
ACRO353030050 ACRO353030250
trans-2-Methyl-2-butenal 98%
trans-2-Methyl-2-butenal
Formel: C₅H₈O
MW: 84,12 g/mol
Kokpunkt: 115…117 °C (1013 hPa)
Densitet: 0,871 g/cm³
Flampunkt: 18 °C
MDL Number: MFCD00006977
CAS nummer: 497-03-0
EINECS: 207-833-0
UN: 1989
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥97.5 %