Etylakrylat 99.5% stabilised
Leverantör: Thermo Scientific
Fara
Synonyms: Acrylic acid-ethyl ester
117980010 117980025 117980250 117980100
ACRO117980010EA 895 SEK
ACRO117980010 ACRO117980025 ACRO117980250 ACRO117980100
Etylakrylat 99.5% stabilised
Etylakrylat
Formel: C₅H₈O₂
MW: 100,12 g/mol
Kokpunkt: 99 °C (760 mmHg)
Smältpunkt: –71 °C
Densitet: 0,92 g/cm³
Flampunkt: 16 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00009188
CAS nummer: 140-88-5
EINECS: 205-438-8
UN: 1917
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4050 to 1.4070 (20°C, 589 nm)
Water ≤0.05 % (K.F.)
GC ≥99.40 %
Color scale ≤10 APHA
Stabilizer 10 to 20 ppm MEHQ
Free acid ≤0.009 % (as acrylic acid)
Heavy metals (ICP-OES) ≤0.001 %
Insoluble matter ≤0.01 %