Tributylphenyltin

Leverantör: Thermo Scientific
Fara

Synonyms: Tri-n-butylphenyltin, Tributylphenylstannane

370230010 370230100 370230500
ACRO370230010EA 505 SEK
ACRO370230010 ACRO370230100 ACRO370230500
Tributylphenyltin
Tributylphenyltin
Formel: C₁₈H₃₂Sn
MW: 367,16 g/mol
Kokpunkt: 125…128 °C (,1 mmHg)
Densitet: 1,125 g/cm³
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00134394
CAS nummer: 960-16-7
UN: 2788
ADR: 6.1,III

Order Now

Specifikation testresultat

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5150 to 1.5170 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR