Isoamyl alcohol (mixture of isomers), EMPLURA®, Supelco®
Leverantör: Merck
Varning
Synonyms: Amyl alcohol (mixture of isomers), Pentanol mixture of isomers, Pentanol (mixture of isomers)
M-Clarity™ quality level = MQ200
8.22255.1000 8.22255.2500
8.22255.1000EA 1110 SEK
8.22255.1000 8.22255.2500
Isoamyl alcohol (mixture of isomers), EMPLURA®, Supelco®
Isoamyl alcohol (mixture of isomers)
Formel: C₅H₁₂O
MW: 88,15 g/mol
Kokpunkt: 131 °C (1013 hPa)
Smältpunkt: –117 °C
Densitet: 0,81 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00002977
CAS nummer: 30899-19-5
EINECS: 250-378-8
UN: 1105
ADR: 3,III
Order Now
Specifikation testresultat
Assay isomers (GC, area%) ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C) 0.808 - 0.812
Water (K. F.) ≤ 0.30 %
Identity (IR) passes test