Sodium metaborate tetrahydrate ≥98%
Leverantör: Thermo Fisher Scientific
Varning
A11803.0E A11803.0C A11803.36
A11803.0EEA 3175 SEK
A11803.0E A11803.0C A11803.36
Sodium metaborate tetrahydrate ≥98%
Sodium metaborate tetrahydrate
Purity: 98% CAS No.: 10555-76-7 Molecular Formula: NaBO2.4H2O Formula Weight: 137.86 (65.80anhy) Odour: Odourless Melting Point: ca 53° dec. MDL No.: MFCD00149205
Formel: BNaO₂·4H₂O
MW: 137.86 g/mol
Smältpunkt: ∼ 53 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00149205
CAS nummer: 10555-76-7
EINECS: 231-891-6