Natriumetanolat 96%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Sodium ethylate, Sodium ethanolate
L06230.22 L06230.36 L06230.0P
L06230.22EA 760 SEK
L06230.22 L06230.36 L06230.0P
Natriumetanolat 96%
Natriumetanolat
Purity: 96% CAS No.: 141-52-6 Molecular Formula: NaOCH2CH3 Formula Weight: 68.05 Flash Point: 30°(86°F) Odour: Irritating pH: 13 Melting Point: 300° Beilstein Reference: 3593646 Merck Reference: 14,8613 UN No.: 3206 MDL No.: MFCD00012417
Formel: C₂H₅NaO
Smältpunkt: >300 °C
Densitet: 0,868 g/cm³ (20 °C)
Flampunkt: 30 °C (86 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00012417
CAS nummer: 141-52-6
EINECS: 205-487-5
UN: 3206
ADR: 4.2,II
Merck Index: 14,8613