Natriumetanolat, Sigma-Aldrich®

Leverantör: Merck
Fara

Synonyms: Sodium ethylate, Sodium ethanolate

M-Clarity™ quality level = MQ200

8.20871.0250 8.20871.1000
8.20871.0250EA 661 SEK
8.20871.0250 8.20871.1000
Natriumetanolat, Sigma-Aldrich®
Natriumetanolat
Formel: C₂H₅NaO
Smältpunkt: 260 °C
Densitet: 0,868 g/cm³ (20 °C)
Storage Temperature: Kylskåp
MDL Number: MFCD00012417
CAS nummer: 141-52-6
EINECS: 205-487-5
UN: 3206
ADR: 4.2 (8),II

Order Now

Specifikation testresultat

Solubility (20 °C) (decomposition)
Melting Point 260 °C
Molar Mass 68.05 g/mol
Bulk Density 300 kg/m3
pH 13 (5 g/l, H2O, 20 °C)
Ignition Point 30 - 50 °C
Hazard Class 4.2
Hazard Class Description Spontaneously combustible material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR