Butylmetakrylat stabilised, Sigma-Aldrich®
Leverantör: Merck
Varning
Synonyms: Methacrylic acid butyl ester
M-Clarity™ quality level = MQ200
8.00585.0100 8.00585.0500
8.00585.0100EA 297 SEK
8.00585.0100 8.00585.0500
Butylmetakrylat stabilised, Sigma-Aldrich®
Butyl methacrylate
Formel: C₈H₁₄O₂
MW: 142,2 g/mol
Kokpunkt: 163 °C (1013 hPa)
Smältpunkt: –75 °C
Densitet: 0,8951 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00009444
CAS nummer: 97-88-1
EINECS: 202-615-1
UN: 2227
ADR: 3,III
Order Now
Specifikation testresultat
Solubility 0.36 g/l (25 °C)
Melting Point -75 °C
Molar Mass 142.20 g/mol
Boiling Point 163 °C (1013 hPa)
Vapor Pressure 2.7 hPa (20 °C)
Flash Point 46 °C
Refractive Index 1.424 (20 °C, 589 nm)
Explosion Limit 2.00 - 8.00 %(V)
Density 0.89 g/cm3 (20 °C)
Ignition Point 290 °C