Litiummetanolat ≥98%
Leverantör: Thermo Scientific
Fara
Synonyms: Lithium methanolate, Lithium methylate
44129.14 44129.22
44129.14EA 1800 SEK
44129.14 44129.22
Litiummetanolat ≥98%
Litiummetanolat
Purity: 98+% CAS No.: 865-34-9 Molecular Formula: CH3LiO Formula Weight: 37.98 Physical Form: Powder Sensitivity: Air and Moisture Sensitive Appearance: White Melting Point: 500° dec. Beilstein Reference: 3552389 UN No.: 3206 MDL No.: MFCD00036357
Formel: CH₃LiO
MW: 37,98 g/mol
Kokpunkt: 64,6 °C (1013 hPa)
Smältpunkt: >500 °C (dec.)
Densitet: 0,865 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00036357
CAS nummer: 865-34-9
EINECS: 212-737-7
UN: 3206
ADR: 4.2,II