Lanthanum(III) chloride hydrate ≥99.9% (REO, rare earth oxide basis)
Leverantör: Thermo Scientific
Varning
Synonyms: Lanthanum trichloride hydrate
43120.22 43120.36
43120.22EA 1675 SEK
43120.22 43120.36
Lanthanum(III) chloride hydrate ≥99.9% (REO, rare earth oxide basis)
Lanthanum(III) chloride hydrate
Purity: 99.9% CAS No.: 20211-76-1 Molecular Formula: LaCl3.xH2O Formula Weight: 245.27(anhy) Physical Form: Crystalline Odour: Odourless pH: 5 Melting Point: 91° dec. Merck Reference: 14,5363 MDL No.: MFCD00149753
Formel: Cl₃La·H₂O
MW: 263,28 g/mol
Kokpunkt: 1000 °C (1013 hPa)
Smältpunkt: 91 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00149753
CAS nummer: 20211-76-1
EINECS: 233-237-5
Merck Index: 14,5363