Lanthanum(III) chloride hydrate 64,5 - 70%
Leverantör: Thermo Fisher Scientific
Fara
199151000 199155000 199150025
ACRO199151000EA 1200 SEK
ACRO199151000 ACRO199155000 ACRO199150025
Lanthanum(III) chloride hydrate 64,5 - 70%
Lanthanum(III) chloride hydrate
Formel: Cl₃La·H₂O
Kokpunkt: 1000 °C (1013 hPa)
Smältpunkt: 96 °C
Storage Temperature: Omgivande
MDL Number: MFCD00149753
CAS nummer: 20211-76-1
EINECS: 233-237-5
UN: 1759
ADR: 8,III
Order Now
Specifikation testresultat
Appearance White Adhering crystals
Gravimetry 64.5 to 70 %