Dipotassium hexachloroosmate (≥38,7% Os)
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Potassium hexachloroosmate(IV)
12177.01 12177.03 12177.06
12177.01EA 2050 SEK
12177.01 12177.03 12177.06
Dipotassium hexachloroosmate (≥38,7% Os)
Dipotassium hexachloroosmate
CAS No.: 16871-60-6 Molecular Formula: K2Cl6Os Formula Weight: 481.12 Physical Form: Powder Sensitivity: Hygroscopic Appearance: Dark red Melting Point: dec. Solubility: Soluble in water. Sparingly soluble in alcohol Merck Reference: 14,7635 MDL No.: MFCD00011371
Formel: Cl₆K₂Os
MW: 481.14 g/mol
Smältpunkt: 600 Dec.
Densitet: 3.42 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011371
CAS nummer: 16871-60-6
EINECS: 240-893-6