Triethylammonium dihydrogentrifluoride (∼37% HF)
Leverantör: Thermo Scientific
Fara
Synonyms: Triethylamine trihydrofluoride, Triethylamine trishydrofluoride
L14417.06 L14417.14 L14417.22
L14417.06EA 655 SEK
L14417.06 L14417.14 L14417.22
Triethylammonium dihydrogentrifluoride (∼37% HF)
Triethylammonium dihydrogentrifluoride
CAS No.: 73602-61-6 Molecular Formula: C6H15N.3HF Formula Weight: 161.21 Flash Point: 87°(188°F) Refractive Index: 1.3915 Melting Point: -29° Boiling Point: 70°/15mm Beilstein Reference: 5522945 UN No.: 1790 MDL No.: MFCD00043294
Formel: C₆H₁₈F₃N
MW: 161,21 g/mol
Kokpunkt: 70 °C (15 mmHg)
Smältpunkt: –29 °C
Densitet: 0,990
Flampunkt: 87 °C (188 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00043294
CAS nummer: 73602-61-6
EINECS: 277-550-5
UN: 1790
ADR: 8,II