Cerium triacetate sesquihydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Leverantör: Thermo Scientific
Fara
Synonyms: Cerium (III) acetate sesquihydrate, Acetic acid cerium (III) salt sesquihydrate
11324.09 11324.18 11324.30
11324.09EA 1275 SEK
11324.09 11324.18 11324.30
Cerium triacetate sesquihydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Cerium triacetate sesquihydrate
Cerium(III) acetate is used in combination with bromide ion, catalyzes the liquid-phase auto-oxidation of cresols. It is used in glass manufacturing, glass Polishing, optical Glass making and polishing, pigments, pharma, Speciality chemicals electronic, oaint and driers, dyes and pigments, paper industries etc.
Purity: 99.99% CAS No.: 537-00-8 Molecular Formula: Ce(C2H3O2)3.1.5H2O Formula Weight: 344.28 (317.26anhy) Physical Form: Crystalline Aggregates Melting Point: 308° dec. MDL No.: MFCD00051060
Formel: Ce(C₂H₃O₂)₃·1.5H₂O
MW: 344,28 g/mol (317,26 anhy)
Smältpunkt: 308 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00051060
CAS nummer: 537-00-8
EINECS: 208-654-0