Bis(8-hydroxyquinolinium)sulphate monohydrate 98%
Leverantör: Thermo Fisher Scientific
Varning
148451000 148455000 148450025
ACRO148451000EA 915 SEK
ACRO148451000 ACRO148455000 ACRO148450025
Bis(8-hydroxyquinolinium)sulphate monohydrate 98%
Bis(8-hydroxyquinolinium)sulphate monohydrate
Formel: C₁₈H₁₆N₂O₆S·1H₂O
Smältpunkt: 182 °C
Storage Temperature: Omgivande
MDL Number: MFCD00149610
CAS nummer: 207386-91-2
EINECS: 205-137-1
Order Now
Specifikation testresultat
Appearance Yellow Crystalline powder and/or lumps
Infrared spectrum Conforms
Water ≤4.5 %
Titration with CHA ≥97.5 %