Cyclopropyltriphenylphosphonium bromide ≥98%
Leverantör: Thermo Fisher Scientific
Varning
B25102.06 B25102.14 B25102.22
B25102.06EA 875 SEK
B25102.06 B25102.14 B25102.22
Cyclopropyltriphenylphosphonium bromide ≥98%
Cyclopropyltriphenylphosphonium bromide
Purity: 98% CAS No.: 14114-05-7 Molecular Formula: C21H20BrP Formula Weight: 383.27 Melting Point: 183-186° Beilstein Reference: 6413483
Formel: C₂₁H₂₀BrP
MW: 383.27 g/mol
Smältpunkt: 183…186 °C
Storage Temperature: Omgivande
MDL Number: MFCD00011872
CAS nummer: 14114-05-7
EINECS: 237-970-1