Norcamphor 99%
Leverantör: Thermo Scientific
Varning
Synonyms: 2-Oxonorbornane, Bicyclo[2.2.1]heptan-2-one, Bicyclo[2.2.1]-2-heptanone, 2,5-Methanocyclohexanone, 2-Norbornanone
129250050 129250250 129251000
ACRO129250050EA 610 SEK
ACRO129250050 ACRO129250250 ACRO129251000
Norcamphor 99%
Norcamphor
Formel: C₇H₁₀O
MW: 110,16 g/mol
Kokpunkt: 168…172 °C (760 mmHg)
Smältpunkt: 88…91 °C
Densitet: 1,03 g/cm³ (25 °C)
Flampunkt: 33 °C
Storage Temperature: Omgivande
MDL Number: MFCD00074823
CAS nummer: 497-38-1
EINECS: 207-846-1
UN: 1325
ADR: 4.1,II
Order Now
Specifikation testresultat
Appearance Colorless to white Adhering crystals
Infrared spectrum Conforms
GC ≥98.5 %
UV at 320 nm A: ≤0.03
Color scale ≤10 APHA
Residue after evaporation ≤5 ppm
Titration with NaOH ≥99.0 %