Daidzein ≥97%
Leverantör: Thermo Scientific
Varning
Synonyms: Daidzeol, 7-Hydroxy-3-(4-hydroxyphenyl)-4-benzopyrone, 7-Hydroxy-3-(4-hydroxyphenyl)chromen-4-one, 4',7-Dihydroxyisoflavone, 7-Hydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one
B22877.03 B22877.06 B22877.14
B22877.03EA 1350 SEK
B22877.03 B22877.06 B22877.14
Daidzein ≥97%
Daidzein
Purity: 97% CAS No.: 486-66-8 Molecular Formula: C15H10O4 Formula Weight: 254.24 Melting Point: 328-332° dec. Beilstein Reference: 231523 Merck Reference: 14,2801 MDL No.: MFCD00016954
Formel: C₁₅H₁₀O₄
MW: 254,24 g/mol
Kokpunkt: 513 °C (1013 hPa)
Smältpunkt: 328…332 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00016954
CAS nummer: 486-66-8
EINECS: 207-635-4
Merck Index: 14,2801