Daidzein 98%
Leverantör: Thermo Scientific
Varning
Synonyms: Daidzeol, 7-Hydroxy-3-(4-hydroxyphenyl)-4-benzopyrone, 7-Hydroxy-3-(4-hydroxyphenyl)chromen-4-one, 4',7-Dihydroxyisoflavone, 7-Hydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one
328230250 328231000
ACRO328230250EA 1450 SEK
ACRO328230250 ACRO328231000
Daidzein 98%
Daidzein
Formel: C₁₅H₁₀O₄
MW: 254,24 g/mol
Kokpunkt: 513 °C (1013 hPa)
Smältpunkt: 212 - 214 °C
Storage Temperature: Frys
MDL Number: MFCD00016954
CAS nummer: 486-66-8
EINECS: 207-635-4
Order Now
Specifikation testresultat
Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥97.5 %