4-Vinylpyridine 95% stabilised
Leverantör: Thermo Fisher Scientific
Fara
140912500 140910010 140911000
ACRO140912500EA 1575 SEK
ACRO140912500 ACRO140910010 ACRO140911000
4-Vinylpyridine 95% stabilised
4-Vinylpyridine
Formel: C₇H₇N
Kokpunkt: 62…65 °C (15 mmHg)
Smältpunkt: –20 °C
Densitet: 0,989 g/cm³
Flampunkt: 48 °C
Storage Temperature: Frys
MDL Number: MFCD00006447
CAS nummer: 100-43-6
EINECS: 202-852-0
UN: 3073
ADR: 6.1,II
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow to red-brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5480 to 1.5540 (20°C, 589 nm)
GC ≥94.0 %
Stabilizer 80 to 150 ppm Hydroquinone