4,5-Dichloroimidazole ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: 4,5-Dichloro-1H-imidazole
L06262.06 L06262.14
L06262.06EA 1125 SEK
L06262.06 L06262.14
4,5-Dichloroimidazole ≥98%
4,5-Dichloroimidazole
Purity: 98% CAS No.: 15965-30-7 Molecular Formula: C3H2Cl2N2 Formula Weight: 136.97 Melting Point: 179-182° Beilstein Reference: 606155
Formel: C₃H₂Cl₂N₂
MW: 136,97 g/mol
Smältpunkt: 179…182 °C
Storage Temperature: Omgivande
MDL Number: MFCD00005195
CAS nummer: 15965-30-7