Ethyl-4,4,4-trifluoroacetoacetate 97%
Leverantör: Thermo Scientific
Varning
Synonyms: Ethyl trifluoroacetoacetate, Ethyl 4,4,4-trifluoro-3-oxobutanoate
119030250 119031000
ACRO119030250EA 1225 SEK
ACRO119030250 ACRO119031000
Ethyl-4,4,4-trifluoroacetoacetate 97%
Ethyl-4,4,4-trifluoroacetoacetate
Formel: C₆H₇F₃O₃
MW: 184,11 g/mol
Kokpunkt: 131 °C (709 mmHg)
Smältpunkt: –39 °C
Densitet: 1,25 g/cm³
Flampunkt: 28 °C
MDL Number: MFCD00000424
CAS nummer: 372-31-6
EINECS: 206-750-7
UN: 1993
ADR: 3,III
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3750 to 1.3780 (20°C, 589 nm)
GC ≥96.0 %