Methyl-3-phenylpropionate ≥98%
Leverantör: Thermo Scientific
Synonyms: Methyl dihydrocinnamate, 3-Phenylpropionic acid methyl ester, Methyl 3-phenylpropionate, Hydrocinnamic acid methyl ester
L05310.18 L05310.30
L05310.18EA 2425 SEK
L05310.18 L05310.30
Methyl-3-phenylpropionate ≥98%
Methyl-3-phenylpropionate
Purity: 98% CAS No.: 103-25-3 Molecular Formula: C10H12O2 Formula Weight: 164.20 Flash Point: 100°(212°F) Refractive Index: 1.5020 Boiling Point: 236-239° Beilstein Reference: 1909591 MDL No.: MFCD00017209
Formel: C₁₀H₁₂O₂
MW: 164,2 g/mol
Kokpunkt: 236…239 °C
Densitet: 1,045
Flampunkt: 100 °C (212 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00017209
CAS nummer: 103-25-3
EINECS: 203-092-2