3-Phenylpropionyl chloride 98%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Hydrocinnamoyl chloride, Hydrocinnamoyl chlorid, 3-Phenylpropanoyl chloride, 3-Phenylpropanonylchlorid
134380250 134381000
ACRO134380250EA 900 SEK
ACRO134380250 ACRO134381000
3-Phenylpropionyl chloride 98%
3-Phenylpropionyl chloride
Formel: C₉H₉ClO
Kokpunkt: 107 °C (11 mmHg)
Smältpunkt: –6 °C
Densitet: 1,137 g/cm³
Flampunkt: 108 °C
MDL Number: MFCD00000748
CAS nummer: 645-45-4
EINECS: 211-443-6
UN: 3265
ADR: 8,II
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5245 to 1.5285 (20°C, 589 nm)
GC ≥97.5 %