3-Nitrothiobenzamide ≥97%
Leverantör: Thermo Fisher Scientific
Varning
H52152.03 H52152.06
H52152.03EA 1200 SEK
H52152.03 H52152.06
3-Nitrothiobenzamide ≥97%
3-Nitrothiobenzamide
Purity: 97% CAS No.: 70102-34-0 Molecular Formula: C7H6N2O2S Formula Weight: 182.20 MDL No.: MFCD09757586
Formel: C₇H₆N₂O₂S
Storage Temperature: Omgivande
MDL Number: MFCD09757586
CAS nummer: 70102-34-0