3-Tolualdehyde 98% stabilised
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: m-Tolualdehyde, 3-Methylbenzaldehyde, m-Tolylaldehyde
138990250 138991000
ACRO138990250EA 985 SEK
ACRO138990250 ACRO138991000
3-Tolualdehyde 98% stabilised
3-Tolualdehyde
Formel: C₈H₈O
Kokpunkt: 199 °C (1013 hPa)
Densitet: 1,019 g/cm³
Flampunkt: 78 °C
Storage Temperature: Omgivande
MDL Number: MFCD00003374
CAS nummer: 620-23-5
EINECS: 210-632-0
Order Now
Specifikation testresultat
Appearance Clear yellow to brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5400 to 1.5420 (20°C, 589 nm)
GC ≥97.5 %
Stabilizer 0.1 % hydroquinone