3-Tolualdehyde ≥97% stabilised
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: m-Tolualdehyde, 3-Methylbenzaldehyde, m-Tolylaldehyde
Stabilization: med hydrokinon
A15313.14 A15313.06 A15313.22
A15313.14EA 1375 SEK
A15313.14 A15313.06 A15313.22
3-Tolualdehyde ≥97% stabilised
3-Tolualdehyde
Stabilized with 0.1% hydroquinone
Purity: 97% CAS No.: 620-23-5 Molecular Formula: C8H8O Formula Weight: 120.15 Flash Point: 78°(172°F) Refractive Index: 1.5410 Odour: Almond-like Boiling Point: 198-200° Beilstein Reference: 741964 MDL No.: MFCD00003374
Formel: C₈H₈O
Kokpunkt: 198…200 °C
Densitet: 1,020
Flampunkt: 78 °C (172 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00003374
CAS nummer: 620-23-5
EINECS: 210-632-0