3-Hydroxyflavone ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: 3-Hydroxy-2-phenyl-4H-benzo[b]pyran-4-one, 3-Hydroxy-2-phenyl-4H-chromen-4-one, 3-Hydroxy-2-phenyl-4-benzopyrone, 3-Hydroxy-2-phenylchromen-4-one, Flavonol, 3-Hydroxy-2-phenyl-4H-1-benzopyran-4-one
A18110.03 A18110.06
A18110.03EA 755 SEK
A18110.03 A18110.06
3-Hydroxyflavone ≥98%
3-Hydroxyflavone
Purity: 98+% CAS No.: 577-85-5 Molecular Formula: C15H10O3 Formula Weight: 238.25 Melting Point: 168-172° Beilstein Reference: 15789
Formel: C₁₅H₁₀O₃
MW: 238,24 g/mol
Kokpunkt: 411 °C (1013 hPa)
Smältpunkt: 168…172 °C
Densitet: 1,393 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00006832
CAS nummer: 577-85-5
EINECS: 209-416-9