2-Klor-p-toluensulfoklorid

Leverantör: Thermo Scientific

Synonyms: 3-Chlor-4-methylbenzenesulphonyl chloride, 2-Chloro-p-toluenesulphochloride, 3-Chloro-4-methylbenzene-1-sulphonyl chloride

TL00685DA TL00685EA
MAYBTL00685DAEA 0 SEK
MAYBTL00685DA MAYBTL00685EA
2-Klor-p-toluensulfoklorid
2-Klor-p-toluensulfoklorid
Formel: C₇H₆Cl₂O₂S
MW: 225,09 g/mol
MDL Number: MFCD00203987
CAS nummer: 42413-03-6
EINECS: 412-890-1
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR