3-Chloro-2-methylbenzenesulphonyl chloride

Leverantör: Thermo Scientific

Synonyms: 3-Chloro-2-methylbenzene-1-sulphonyl chloride

TL00453.1 TL00453EA
MAYBTL00453.1EA 0 SEK
MAYBTL00453.1 MAYBTL00453EA
3-Chloro-2-methylbenzenesulphonyl chloride
3-Chloro-2-methylbenzenesulphonyl chloride
Formel: C₇H₆Cl₂O₂S
MW: 225,09 g/mol
Smältpunkt: 72 °C
MDL Number: MFCD00052198
CAS nummer: 80563-86-6
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR