3-Amino-9-etylkarbazol (AEC) 90% tech.
Leverantör: Thermo Scientific
Fara
Synonyms: 9-Etylkarbazol-3-ylamin
147870050 147870250 147871000
ACRO147870050EA 630 SEK
ACRO147870050 ACRO147870250 ACRO147871000
3-Amino-9-etylkarbazol (AEC) 90% tech.
3-Amino-9-etylkarbazol (AEC)
Formel: C₁₄H₁₄N₂
MW: 210,28 g/mol
Smältpunkt: 98…100 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004964
CAS nummer: 132-32-1
EINECS: 205-057-7
UN: 2811
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance Brown Crystalline powder and/or chunks
Infrared spectrum Conforms
GC ≥88 %