3,5-Difluoro-4-hydroxybenzaldehyde 97%
Leverantör: Thermo Scientific
Varning
H26139.MD H26139.03
H26139.MDEA 2400 SEK
H26139.MD H26139.03
3,5-Difluoro-4-hydroxybenzaldehyde 97%
3,5-Difluoro-4-hydroxybenzaldehyde
Purity: 97% CAS No.: 118276-06-5 Molecular Formula: C7H4F2O2 Formula Weight: 158.11 Melting Point: 121-124° MDL No.: MFCD06797917
Formel: C₇H₄F₂O₂
Smältpunkt: 121…124 °C
Storage Temperature: Omgivande
MDL Number: MFCD06797917
CAS nummer: 118276-06-5