3,5-Bis(trifluoromethyl)benzenesulphonyl chloride ≥97%
Leverantör: Thermo Scientific
Fara
Synonyms: 3,5-Bis(trifluoromethyl)benzene-1-sulphonyl chloride
A10300.03 A10300.06
A10300.03EA 1275 SEK
A10300.03 A10300.06
3,5-Bis(trifluoromethyl)benzenesulphonyl chloride ≥97%
3,5-Bis(trifluoromethyl)benzenesulphonyl chloride
Purity: 97% CAS No.: 39234-86-1 Molecular Formula: C8H3ClF6O2S Formula Weight: 312.61 Melting Point: 34-38° Beilstein Reference: 3621218 UN No.: 3261 MDL No.: MFCD00014725
Formel: C₈H₃ClF₆O₂S
MW: 312,62 g/mol
Smältpunkt: 34…38 °C
Storage Temperature: Omgivande
MDL Number: MFCD00014725
CAS nummer: 39234-86-1
EINECS: 254-371-0
UN: 3261
ADR: 8,II